Learn More
SAI Infusion Technologies Sterile Press'n Seal - 140 sq feet
SDP
Catalog No. NC0858581
Encompass
Press'n Seal - 140 sq feet ETO Exposed for Sterility. Provides a transparent barrier for patient monitoring in rodent surgeries. Product retains body heat, hair, and allows for easy repositioning during surgery. For use as sterile surgical drape in preclinical studies. 140' roll (70' also available) Product is for laboratory animal research only.
Catalog No. NC0858581 Supplier SAI Infusion Technologies Supplier No. PSS140
Press'n Seal - 140 sq feet ETO Exposed for Sterility. Provides a transparent barrier for patient monitoring in rodent surgeries. Product retains body heat, hair, and allows for easy repositioning during surgery. For use as sterile surgical drape in preclinical studies. 140' roll (70' also available) Product is for laboratory animal research only.