Learn More
AdipoGen Etobenzanid
SDP
Supplier:  AdipoGen CDXE0065M025
Encompass_Preferred
Chemical. CAS 79540-50-4. Formula C16H15Cl2NO3. MW 340.2. Synthetic. Herbicide. Compound can be used as analytical reference material.
Catalog No. 50-256-0980