Learn More
Apexbio Technology LLC Etodolac 41340-25-4 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061747
Encompass_Preferred
Etodolac (CAS 41340-25-4) is a selective cyclooxygenase-2 (COX-2) inhibitor belonging to the class of non-steroidal anti-inflammatory drugs (NSAIDs) It exhibits inhibitory activity against COX-2 with an IC50 of 53 5 nM Etodolac is commonly utilized for research involving inflammatory conditions such as osteoarthritis and rheumatoid arthritis In isolated rabbit articular chondrocytes stimulated with TNF Etodolac prevented apoptotic cell shrinkage and suppressed caspase-3/7 activation Conversely in human malignant rhabdoid tumor cells (FRTK-1) Etodolac induced apoptosis in a dose-dependent manner increasing activities of caspase-3 -8 and -9
Catalog No. 50-000-61747 Supplier Apexbio Technology LLC Supplier No. B14445.1
Etodolac (CAS 41340-25-4) is a selective cyclooxygenase-2 (COX-2) inhibitor belonging to the class of non-steroidal anti-inflammatory drugs (NSAIDs) It exhibits inhibitory activity against COX-2 with an IC50 of 53 5 nM Etodolac is commonly utilized for research involving inflammatory conditions such as osteoarthritis and rheumatoid arthritis In isolated rabbit articular chondrocytes stimulated with TNF Etodolac prevented apoptotic cell shrinkage and suppressed caspase-3/7 activation Conversely in human malignant rhabdoid tumor cells (FRTK-1) Etodolac induced apoptosis in a dose-dependent manner increasing activities of caspase-3 -8 and -9