Learn More
Cayman Chemical Etodolac Acyl Glucuronide, Purity: ≥90%, CAS: 79541-43-8, 1 mg
SDP
Catalog No. 502950517
Encompass_Preferred
A phase II metabolite of etodolac; formed via glucuronidation of etodolac by UGT1A9, UGT1A10, and UGT2B7
Catalog No. 50-295-0517 Supplier Cayman Chemical Supplier No. 340721 MG
A phase II metabolite of etodolac; formed via glucuronidation of etodolac by UGT1A9, UGT1A10, and UGT2B7