Learn More
Medchemexpress LLC HY-B1209 500mg Medchemexpress, Etofylline CAS:519-37-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB1209500MG
Encompass_Preferred
Medchemexpress, HY-B1209 500mg Etofylline CAS:519-37-9 Etofylline (7-(β-Hydroxyethyl)theophylline) is a N-7-substituted derivative of Theophylline. Etofylline is a bronchodilator which can be used for the research of asthma. Etofylline is also an anticholesteremic and reduces total cholesterol level in the blood. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2692