Learn More
Medchemexpress LLC Etofylline clofibrate | 54504-70-0 | MFCD00865733 | 99.8% | 420.85 g/mol | C19H21ClN4O5 | 10 MG
SDP
Catalog No. 5000239697
Encompass_Preferred
Etofylline clofibrate is a small-molecule research compound reported to have hypolipidemic and antithrombotic effects, including an agonistic interaction with intimal prostacyclin (PGI2). It is supplied as a high-purity analytical material intended for laboratory research and is not for human or veterinary use.
  • Hypolipidemic and antithrombotic pharmacological profile.
  • Agonistic interaction with intimal prostacyclin (PGI2).
  • High purity for analytical and research applications (~99.8%).
  • Molecular weight 420.85 g/mol; formula C19H21ClN4O5.
  • Supplied in small-scale quantities suitable for in vitro and analytical work.
Catalog No. 50-002-39697 Supplier Medchemexpress LLC Supplier No. HY10734810MG
Etofylline clofibrate is a small-molecule research compound reported to have hypolipidemic and antithrombotic effects, including an agonistic interaction with intimal prostacyclin (PGI2). It is supplied as a high-purity analytical material intended for laboratory research and is not for human or veterinary use.
  • Hypolipidemic and antithrombotic pharmacological profile.
  • Agonistic interaction with intimal prostacyclin (PGI2).
  • High purity for analytical and research applications (~99.8%).
  • Molecular weight 420.85 g/mol; formula C19H21ClN4O5.
  • Supplied in small-scale quantities suitable for in vitro and analytical work.