Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Etofylline clofibrate | 54504-70-0 | MFCD00865733 | 99.8% | 420.85 g/mol | C19H21ClN4O5 | 100 MG
SDP

Catalog No. 5000239813
Encompass_Preferred

Etofylline clofibrate is a research compound reported to have hypolipidemic and antithrombotic effects, including an agonistic interaction with intimal PGI2. It is supplied as a purified solid and in solution formats for use in in vitro and in vivo pharmacology studies, with documented solubility and storage recommendations.

  • Reported hypolipidemic and antithrombotic activity with agonistic effects on intimal PGI2.
  • High purity (≈99.8%) suitable for research applications.
  • Molecular formula C19H21ClN4O5 and molecular weight 420.85 g/mol.
  • Soluble in DMSO (≈52 mg/mL); in vivo formulation protocols available to achieve ≥2.17 mg/mL.
  • Stable as a powder at -20°C for long-term storage; in solution store at -80°C for extended stability.
  • Available as small solid quantities and ready-to-use solution formats for immediate experimentation.

Catalog No. 50-002-39813 Supplier Medchemexpress LLC Supplier No. HY107348100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Etofylline clofibrate is a research compound reported to have hypolipidemic and antithrombotic effects, including an agonistic interaction with intimal PGI2. It is supplied as a purified solid and in solution formats for use in in vitro and in vivo pharmacology studies, with documented solubility and storage recommendations.

  • Reported hypolipidemic and antithrombotic activity with agonistic effects on intimal PGI2.
  • High purity (≈99.8%) suitable for research applications.
  • Molecular formula C19H21ClN4O5 and molecular weight 420.85 g/mol.
  • Soluble in DMSO (≈52 mg/mL); in vivo formulation protocols available to achieve ≥2.17 mg/mL.
  • Stable as a powder at -20°C for long-term storage; in solution store at -80°C for extended stability.
  • Available as small solid quantities and ready-to-use solution formats for immediate experimentation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.