Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Etofylline clofibrate | 54504-70-0 | MFCD00865733 | 99.8% | 420.85 g/mol | C19H21ClN4O5 | 50 MG
SDP

Catalog No. 5000239784
Encompass_Preferred

Etofylline clofibrate is a small-molecule research reagent reported to have hypolipidemic and antithrombotic activity, including an agonistic interaction with intimal prostacyclin (PGI2). It is supplied as a high-purity analytical standard for biochemical and pharmacological research and assay development.

  • High purity: 99.77%.
  • Molecular weight: 420.85 g/mol.
  • Molecular formula: C19H21ClN4O5.
  • Intended for research use and analytical applications.
  • Available in small-mass pack sizes suitable for assay development.

Catalog No. 50-002-39784 Supplier Medchemexpress LLC Supplier No. HY10734850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Etofylline clofibrate is a small-molecule research reagent reported to have hypolipidemic and antithrombotic activity, including an agonistic interaction with intimal prostacyclin (PGI2). It is supplied as a high-purity analytical standard for biochemical and pharmacological research and assay development.

  • High purity: 99.77%.
  • Molecular weight: 420.85 g/mol.
  • Molecular formula: C19H21ClN4O5.
  • Intended for research use and analytical applications.
  • Available in small-mass pack sizes suitable for assay development.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.