Learn More
Apexbio Technology LLC Etomidate, 25mg. Cas: 33125-97-2 MFCD: MFCD00867535. General anesthetic with GABA modulatory and GABA-mimetic actions
SDP
Supplier:  Apexbio Technology LLC A195825
Encompass_Preferred
General anesthetic with GABA modulatory and GABA-mimetic actions; selectively interacts with 2- and 3-subunit containing GABAA receptors. Short acting and potent hypnotic, with low toxicity. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4729
 Restricted Item