Learn More
Medchemexpress LLC HY-50202 10mg Medchemexpress, Etomoxir CAS:124083-20-1 Purity:>98%
SDP
Catalog No. 502029321
Encompass_Preferred
Medchemexpress, HY-50202 10mg Etomoxir CAS:124083-20-1 Etomoxir ((R)-(+)-Etomoxir) is an irreversible inhibitor of carnitine palmitoyltransferase 1a (CPT-1a), inhibits fatty acid oxidation (FAO) through CPT-1a and inhibits palmitate β-oxidation in human, rat and guinea pig. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9321 Supplier Medchemexpress LLC Supplier No. HY5020210MG
Medchemexpress, HY-50202 10mg Etomoxir CAS:124083-20-1 Etomoxir ((R)-(+)-Etomoxir) is an irreversible inhibitor of carnitine palmitoyltransferase 1a (CPT-1a), inhibits fatty acid oxidation (FAO) through CPT-1a and inhibits palmitate β-oxidation in human, rat and guinea pig. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.