Learn More
Apexbio Technology LLC Etomoxir(Synonyms: Etomoxir sodium salt, Ethyl 2-[6-(4-chlorophenoxy)hexyl]oxirane-2-carboxylate), 10mg, CAS: 124083-20-1.
SDP
Supplier:  Apexbio Technology LLC A340410
Encompass_Preferred
Etomoxir (CAS 124083-20-1) is a cell-permeable small molecule known to act as an irreversible and stereospecific inhibitor of mitochondrial carnitine palmitoyltransferase-1 (CPT-1) Additionally it has demonstrated inhibitory activity against diacylglycerol acyltransferase (DGAT) In rat heart-derived H9c2 myoblastic cells CPT-1 inhibition occurs at concentrations between 1 to 80 M whereas DGAT activity is inhibited at around 40 M Etomoxir is utilized in research to investigate fatty acid oxidation pathways and related metabolic disorders
Catalog No. 50-223-9980