Learn More
Medchemexpress LLC HY-50202 50mg , Etomoxir CAS:124083-20-1 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY5020250MG
Encompass_Preferred
Medchemexpress, HY-50202 50mg Etomoxir CAS:124083-20-1 Formula:C17H23ClO4 CPT-1 Purity:98% Etomoxir is a potent inhibitor of carnitine palmitoyltransferase-I ( CPT-1 ). Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3619