Learn More
Selleck Chemical LLC Etomoxir sodium salt
SDP
Catalog No. 5000001724
Encompass_Preferred
Etomoxir sodium salt ((R)-( )-Etomoxir sodium salt) is an irreversible inhibitor of carnitine palmitoyltransferase-1 (CPT-1) on the outer face of the inner mitochondrial membrane Etomoxir enhances palmitate-induced cell apoptosis
Catalog No. 50-000-01724 Supplier Selleck Chemical LLC Supplier No. S8244100MG
Etomoxir sodium salt ((R)-( )-Etomoxir sodium salt) is an irreversible inhibitor of carnitine palmitoyltransferase-1 (CPT-1) on the outer face of the inner mitochondrial membrane Etomoxir enhances palmitate-induced cell apoptosis