Learn More
Cayman Chemical (+)-Etomoxir (sodium salt), 828934-41-4, 50 mg
SDP
Supplier:  Cayman Chemical 1196950
Encompass_Preferred
An irreversible inhibitor of carnitine palmitoyltransferase 1, a mitochondrial enzyme involved in fatty acid β-oxidation (IC50 = 5-20 nM in rat liver); also acts as a direct agonist of PPARα
Catalog No. NC1029146
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.