Learn More
Medchemexpress LLC Etomoxir sodium salt | 828934-41-4 | MFCD07787411 | >98.0% | 320.74 g/mol | C15H18ClNaO4 | 10 MG
SDP
Catalog No. 5000298454
Encompass_Preferred
Etomoxir sodium salt is the sodium salt form of etomoxir, an irreversible inhibitor of carnitine palmitoyltransferase 1a (CPT1a) used in research to inhibit fatty acid oxidation and study metabolic pathways. Supplied as a solid suitable for laboratory experiments.
  • Irreversible CPT1a inhibitor for fatty acid oxidation studies
  • High purity suitable for research applications
  • Sodium salt form improves handling and solubility in polar solvents
  • Used in metabolic, apoptosis, and mitochondrial research
  • Typically stored at low temperature to maintain stability
Catalog No. 50-002-98454 Supplier Medchemexpress LLC Supplier No. HY50202A10MG
Etomoxir sodium salt is the sodium salt form of etomoxir, an irreversible inhibitor of carnitine palmitoyltransferase 1a (CPT1a) used in research to inhibit fatty acid oxidation and study metabolic pathways. Supplied as a solid suitable for laboratory experiments.
  • Irreversible CPT1a inhibitor for fatty acid oxidation studies
  • High purity suitable for research applications
  • Sodium salt form improves handling and solubility in polar solvents
  • Used in metabolic, apoptosis, and mitochondrial research
  • Typically stored at low temperature to maintain stability