Learn More
Cayman Chemical (+)-Etomoxir (sodium salt), Purity: ≥98%, CAS: 828934-41-4, 10 mg
SDP
Catalog No. 502954090
Encompass_Preferred
An inhibitor of CPT1; inhibits fatty acid transport into the mitochondria for β-oxidation; inhibits fatty acid oxidation in hepatocytes in vitro (IC50s = 0.1, 1, and 10 µM for human, guinea pig, and rat hepatocytes, respectively); decreases ATP-linked and uncoupled oxygen consumption in C2C12 myotubes when used in combination with BPTES or UK 5099
Catalog No. 50-295-4090 Supplier Cayman Chemical Supplier No. 1196910 MG
An inhibitor of CPT1; inhibits fatty acid transport into the mitochondria for β-oxidation; inhibits fatty acid oxidation in hepatocytes in vitro (IC50s = 0.1, 1, and 10 µM for human, guinea pig, and rat hepatocytes, respectively); decreases ATP-linked and uncoupled oxygen consumption in C2C12 myotubes when used in combination with BPTES or UK 5099