Learn More
Cayman Chemical Etongestrel 10mg
SDP
Catalog No. 502982302
Encompass_Preferred
A progesterone and androgen receptor antagonist (IC50s = 0.51 and 0.12 nM for isolated rabbit uterus and rat prostate cells, respectively) and an active metabolite of desogestrel; increases endometrial and myometrial vascularization, uterine weight, and uterine artery length, as well as decreases the number of hormone-secreting corpus luteal glands in non-ovariectomized guinea pigs when administered subcutaneously as a 1 mm sustained-release pellet
Catalog No. 50-298-2302 Supplier Cayman Chemical Supplier No. 2106210 MG
A progesterone and androgen receptor antagonist (IC50s = 0.51 and 0.12 nM for isolated rabbit uterus and rat prostate cells, respectively) and an active metabolite of desogestrel; increases endometrial and myometrial vascularization, uterine weight, and uterine artery length, as well as decreases the number of hormone-secreting corpus luteal glands in non-ovariectomized guinea pigs when administered subcutaneously as a 1 mm sustained-release pellet