Learn More
Apexbio Technology LLC Etonogestrel(Synonyms: 3-Ketodesogestrel, Implanon, Nexplanon, ORG-3236, 11-Methylenelevonorgestrel), 10mM (in 1mL DMSO), CAS: 54048-10-1.
SDP
Catalog No. 5000060650
Encompass_Preferred
Etonogestrel (CAS 54048-10-1) is a synthetic steroidal molecule derived as the biologically active metabolite of the progestin desogestrel Structurally it belongs to the 19-nortestosterone class and acts primarily through potent binding and agonist activity at progesterone receptors Through this receptor binding it modulates various physiological processes including ovulation suppression cervical mucus viscosity enhancement and endometrial alteration Etonogestrel serves as a widely researched hormonal contraceptive with applications in reproductive biology studies endocrinological research and pharmaceutical development
Catalog No. 50-000-60650 Supplier Apexbio Technology LLC Supplier No. B34705.1
Etonogestrel (CAS 54048-10-1) is a synthetic steroidal molecule derived as the biologically active metabolite of the progestin desogestrel Structurally it belongs to the 19-nortestosterone class and acts primarily through potent binding and agonist activity at progesterone receptors Through this receptor binding it modulates various physiological processes including ovulation suppression cervical mucus viscosity enhancement and endometrial alteration Etonogestrel serves as a widely researched hormonal contraceptive with applications in reproductive biology studies endocrinological research and pharmaceutical development