Learn More
Selleck Chemical LLC Etonogestrel 50mg 54048-10-1 Implanon, Nexplanon, 3-Oxodesogestrel, 3-keto-Desogestrel
SDP
Supplier:  Selleck Chemical LLC S467350MG
Encompass_Preferred
Etonogestrel (Implanon, Nexplanon, 3-Oxodesogestrel, 3-keto-Desogestrel) is a synthetic form of the naturally occurring female sex hormone progesterone. It binds to the cytoplasmic progesterone receptors in the reproductive system and subsequently activates progesterone receptor mediated gene expression. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4153