Learn More
Alkali Scientific Etoposide, 1 G
SDP
Catalog No. 5000661009
Encompass_Preferred
Etoposide, 1g, is a chemotherapy drug used in cancer research to evaluate its effects on tumor cells. This compound inhibits the enzyme topoisomerase II, which is crucial for DNA replication. By blocking this enzyme, etoposide induces DNA damage that leads to cell death, especially in rapidly dividing cells like those found in tumors. The 1g dosage is ideal for large-scale experiments or studies that require high concentrations of the drug. Researchers use this formulation in cancer studies to understand the mechanisms of DNA damage and cell death, as well as to test its efficacy in combination with other chemotherapeutic agents.
Catalog No. 50-006-61009 Supplier Alkali Scientific Supplier No. ETO004CP
Etoposide, 1g, is a chemotherapy drug used in cancer research to evaluate its effects on tumor cells. This compound inhibits the enzyme topoisomerase II, which is crucial for DNA replication. By blocking this enzyme, etoposide induces DNA damage that leads to cell death, especially in rapidly dividing cells like those found in tumors. The 1g dosage is ideal for large-scale experiments or studies that require high concentrations of the drug. Researchers use this formulation in cancer studies to understand the mechanisms of DNA damage and cell death, as well as to test its efficacy in combination with other chemotherapeutic agents.