Learn More
Alkali Scientific Etoposide, 100 Mg
SDP
Catalog No. 5000660382
Encompass_Preferred
Etoposide, 100mg, is a chemotherapy drug used in laboratory studies to explore the effects of cancer treatments on tumor cells. By inhibiting topoisomerase II, Etoposide induces DNA damage, which triggers cell death, particularly in rapidly dividing cancer cells. This potent anticancer agent is often used in experiments testing its effects on various cancer cell lines and its potential as part of combination therapies. The 100mg volume is suitable for larger-scale studies, offering an efficient amount for detailed investigation into its mechanism of action, pharmacodynamics, and potential therapeutic applications in oncology.
Catalog No. 50-006-60382 Supplier Alkali Scientific Supplier No. ETO002CP
Etoposide, 100mg, is a chemotherapy drug used in laboratory studies to explore the effects of cancer treatments on tumor cells. By inhibiting topoisomerase II, Etoposide induces DNA damage, which triggers cell death, particularly in rapidly dividing cancer cells. This potent anticancer agent is often used in experiments testing its effects on various cancer cell lines and its potential as part of combination therapies. The 100mg volume is suitable for larger-scale studies, offering an efficient amount for detailed investigation into its mechanism of action, pharmacodynamics, and potential therapeutic applications in oncology.