Learn More
Apexbio Technology LLC Etoposide, 33419-42-0, MFCD00869325, 100 mg
SDP
Catalog No. 501011642
Encompass_Preferred
MF: C29H32O13, MW: 588.56, Solubility: 29.45mg/mL in DMSO. Etoposide (VP-16) is the first agent recognized as a topoisomerase II inhibitor of anticancer drug with IC50 of 59.2 uM.
Catalog No. 50-101-1642 Supplier Apexbio Technology LLC Supplier No. A1971100
MF: C29H32O13, MW: 588.56, Solubility: 29.45mg/mL in DMSO. Etoposide (VP-16) is the first agent recognized as a topoisomerase II inhibitor of anticancer drug with IC50 of 59.2 uM.