Learn More
Apexbio Technology LLC Etoposide 33419-42-0 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061420
Encompass_Preferred
Etoposide (VP-16) is a topoisomerase II inhibitor widely used in biomedical research to study mechanisms of DNA damage and cancer therapy It forms a stabilized complex with DNA and topoisomerase II preventing the enzyme-mediated religation of cleaved DNA strands Resulting DNA double-strand breaks lead to apoptosis particularly in rapidly dividing cancer cells Reported IC50 values vary across cell lines such as 59 2 M for topoisomerase II inhibition 30 16 M in HepG2 and 0 051 M for MOLT-3 indicating differential cytotoxic responses to etoposide in various cancer cell models
Catalog No. 50-000-61420 Supplier Apexbio Technology LLC Supplier No. A19715.1
Etoposide (VP-16) is a topoisomerase II inhibitor widely used in biomedical research to study mechanisms of DNA damage and cancer therapy It forms a stabilized complex with DNA and topoisomerase II preventing the enzyme-mediated religation of cleaved DNA strands Resulting DNA double-strand breaks lead to apoptosis particularly in rapidly dividing cancer cells Reported IC50 values vary across cell lines such as 59 2 M for topoisomerase II inhibition 30 16 M in HepG2 and 0 051 M for MOLT-3 indicating differential cytotoxic responses to etoposide in various cancer cell models