Learn More
Medchemexpress LLC HY-13629 200mg Medchemexpress, Etoposide CAS:33419-42-0 Purity:>98%
SDP
Catalog No. 502029012
Encompass_Preferred
Medchemexpress, HY-13629 200mg Etoposide CAS:33419-42-0 Etoposide (VP-16; VP-16-213) is an anti-cancer chemotherapy agent. Etoposide inhibits topoisomerase II, thus stopping DNA replication. Etoposide induces cell cycle arrest, apoptosis and autophagy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9012 Supplier Medchemexpress LLC Supplier No. HY13629200MG
Medchemexpress, HY-13629 200mg Etoposide CAS:33419-42-0 Etoposide (VP-16; VP-16-213) is an anti-cancer chemotherapy agent. Etoposide inhibits topoisomerase II, thus stopping DNA replication. Etoposide induces cell cycle arrest, apoptosis and autophagy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.