Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology Etoposide, Topoisomerase II Inhibitor, >98% Purity, 5.9 mg
SDP

Catalog No. NC1608114
Encompass

Etoposide (VP-16) is an anti-tumor agent that functions as a topoisomerase II inhibitor. It stabilizes the covalent topoisomerase II-cleaved DNA intermediate complex, leading to genomic instability and cell death, and is commonly used as an apoptosis inducer. This chemical modulator is supplied as a lyophilized powder.

  • IC50: 59.2 μM
  • Purity: >98%
  • Mechanism of action: Stabilizes covalent topoisomerase II-cleaved DNA intermediate complex, delaying cell cycle progression through late S and early G2 phases
  • Reconstitution: For a 25 mM stock solution, reconstitute the 5.9 mg in 400 μL DMSO
  • Solubility: 25 mg/mL in DMSO
  • Typical usage: 5–50 μM for 4–24 hr treatments
  • Storage: Store lyophilized or in solution at -20°C, desiccated

Catalog No. NC1608114 Supplier Cell Signaling Technology Supplier No. 2200S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Etoposide (VP-16) is an anti-tumor agent that functions as a topoisomerase II inhibitor. It stabilizes the covalent topoisomerase II-cleaved DNA intermediate complex, leading to genomic instability and cell death, and is commonly used as an apoptosis inducer. This chemical modulator is supplied as a lyophilized powder.

  • IC50: 59.2 μM
  • Purity: >98%
  • Mechanism of action: Stabilizes covalent topoisomerase II-cleaved DNA intermediate complex, delaying cell cycle progression through late S and early G2 phases
  • Reconstitution: For a 25 mM stock solution, reconstitute the 5.9 mg in 400 μL DMSO
  • Solubility: 25 mg/mL in DMSO
  • Typical usage: 5–50 μM for 4–24 hr treatments
  • Storage: Store lyophilized or in solution at -20°C, desiccated

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.