Learn More
Alkali Scientific Etoposide, 25 Mg
SDP
Catalog No. 5000660260
Encompass_Preferred
Etoposide, 25mg, is an anticancer compound used in laboratory research to study the effects of chemotherapy agents. It is commonly used in vitro to examine its ability to inhibit DNA replication and induce apoptosis in cancer cells. Etoposide works by inhibiting topoisomerase II, an enzyme crucial for DNA unwinding during replication. This action leads to DNA strand breaks and ultimately cell death. The 25mg volume is ideal for small-scale laboratory experiments, allowing researchers to test etoposide’s efficacy in various cancer cell lines. It is a crucial reagent in the study of cancer treatment mechanisms and the development of new cancer therapies.
Catalog No. 50-006-60260 Supplier Alkali Scientific Supplier No. ETO001CP
Etoposide, 25mg, is an anticancer compound used in laboratory research to study the effects of chemotherapy agents. It is commonly used in vitro to examine its ability to inhibit DNA replication and induce apoptosis in cancer cells. Etoposide works by inhibiting topoisomerase II, an enzyme crucial for DNA unwinding during replication. This action leads to DNA strand breaks and ultimately cell death. The 25mg volume is ideal for small-scale laboratory experiments, allowing researchers to test etoposide’s efficacy in various cancer cell lines. It is a crucial reagent in the study of cancer treatment mechanisms and the development of new cancer therapies.