Learn More
Alkali Scientific Etoposide, 250 Mg
SDP
Catalog No. 5000659858
Encompass_Preferred
Etoposide, 250mg, is an important anticancer drug used in laboratory experiments focused on cancer cell biology. This compound targets and inhibits topoisomerase II, an enzyme necessary for the unwinding of DNA during replication, causing DNA strand breaks and leading to cell death. Etoposide is widely studied in various experimental cancer therapies and is used in cell lines to assess its cytotoxicity and apoptotic effects. The 250mg formulation provides a higher dosage for more extensive research or clinical trials, making it useful for testing its effects in combination therapies and other cancer treatment strategies.
Catalog No. 50-006-59858 Supplier Alkali Scientific Supplier No. ETO003CP
Etoposide, 250mg, is an important anticancer drug used in laboratory experiments focused on cancer cell biology. This compound targets and inhibits topoisomerase II, an enzyme necessary for the unwinding of DNA during replication, causing DNA strand breaks and leading to cell death. Etoposide is widely studied in various experimental cancer therapies and is used in cell lines to assess its cytotoxicity and apoptotic effects. The 250mg formulation provides a higher dosage for more extensive research or clinical trials, making it useful for testing its effects in combination therapies and other cancer treatment strategies.