Learn More
AdipoGen Etoposide (25 mg)
SDP
Supplier:  AdipoGen AGCR13572M025
Encompass_Preferred
Chemical. CAS: 33419-42-0. Formula: C29H32O13. MW: 588.6. Semisynthetic derivative of podophyllotoxin. Potent anti-cancer compound. Induces apoptosis in normal and tumor cell lines. DNA Topoisomerase II activity inhibitor. Increases Topo II-mediated DNA breakage primarily by inhibiting the ability of the enzyme to religate cleaved nucleic acid molecules. Does not lead to immediate block of DNS synthesis, induces a progressive inhibition of DNA replication. p53 activator. Blocks the cell cycle between the end of the S phase and the early G2 phase. Oncoprotein Mdm2 synthesis inhibitor. Apoptosis inducer through the cytochrome c/Apaf-1/caspase-9 pathway and the Fas-mediated death signaling pathway. Cell cycle checkpoint activator. Affects gene expression at different levels (chromatin remodeling, transcription and alternative splicing). Chemotherapeutic compound used in cancers.
Catalog No. 50-205-3940