Learn More
Medchemexpress LLC Etoposide | 33419-42-0 | 588.56 | 1 ML
SDP
Catalog No. 5000351898
Encompass_Preferred
Etoposide (VP-16; VP-16-213) is an anti-cancer chemotherapy agent that inhibits topoisomerase II, thereby stopping DNA replication and inducing cell cycle arrest, apoptosis, and autophagy.
  • Acts as an anti-cancer chemotherapy agent
  • Inhibits topoisomerase II activity
  • Stops DNA replication
  • Induces cell cycle arrest, apoptosis, and autophagy
  • Soluble in DMSO for in vitro studies
  • Applicable for cancer and infection research
Catalog No. 50-003-51898 Supplier Medchemexpress LLC Supplier No. HY1362910MM/1ML
Etoposide (VP-16; VP-16-213) is an anti-cancer chemotherapy agent that inhibits topoisomerase II, thereby stopping DNA replication and inducing cell cycle arrest, apoptosis, and autophagy.
  • Acts as an anti-cancer chemotherapy agent
  • Inhibits topoisomerase II activity
  • Stops DNA replication
  • Induces cell cycle arrest, apoptosis, and autophagy
  • Soluble in DMSO for in vitro studies
  • Applicable for cancer and infection research