Learn More
Medchemexpress LLC Etoposide | 33419-42-0 | 99.6% | 588.56 | 500 MG
SDP
Supplier:  Medchemexpress LLC 500MGHY13629
Encompass_Preferred
Etoposide (Standard) is the analytical standard of Etoposide, designed for research and analytical applications. It functions as an anti-cancer chemotherapy agent by inhibiting topoisomerase II, which halts DNA replication. This action leads to the induction of cell cycle arrest, apoptosis, and autophagy. As an analytical standard, the compound is frequently utilized in qualitative, quantitative, and methodological research experiments, including techniques such as HPLC, GC, and MS.
  • Inhibits topoisomerase II
  • Halts DNA replication
  • Induces cell cycle arrest, apoptosis, and autophagy
  • Utilized in qualitative, quantitative, and methodological research experiments
  • Suitable for techniques such as HPLC, GC, and MS
Catalog No. 50-003-65914