Learn More
Medchemexpress LLC Etoposide | 33419-42-0 | 99.9% | 588.56 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY13629R
Encompass_Preferred
Etoposide (VP-16; VP-16-213) is an anti-cancer chemotherapy agent that inhibits topoisomerase II, thereby stopping DNA replication. It induces cell cycle arrest, apoptosis, and autophagy, making it a valuable tool in cancer research and related studies.
  • Inhibits topoisomerase II
  • Stops DNA replication
  • Induces cell cycle arrest
  • Promotes apoptosis
  • Triggers autophagy
  • Used as an anti-cancer chemotherapy agent
Catalog No. 50-003-48271