Learn More
Medchemexpress LLC Etoposide | 33419-42-0 | C29H32O13 | 10MG
SDP
Catalog No. 5000261942
Encompass_Preferred
Etoposide (Standard) is an analytical reference standard of the antineoplastic agent etoposide (CAS 33419-42-0). It is intended for research and analytical applications such as assay development, method validation, and quality control. The material has confirmed purity of 99.62% by HPLC and is supplied with supporting documentation for traceability.
  • High purity reference material, 99.62% by HPLC.
  • Intended for assay development and method validation.
  • Suitable for quality control and analytical testing.
  • Applicable to studies of topoisomerase II inhibition and related pharmacology.
  • Provided with a certificate of analysis to support traceability.
Catalog No. 50-002-61942 Supplier Medchemexpress LLC Supplier No. HY13629R10MG
Etoposide (Standard) is an analytical reference standard of the antineoplastic agent etoposide (CAS 33419-42-0). It is intended for research and analytical applications such as assay development, method validation, and quality control. The material has confirmed purity of 99.62% by HPLC and is supplied with supporting documentation for traceability.
  • High purity reference material, 99.62% by HPLC.
  • Intended for assay development and method validation.
  • Suitable for quality control and analytical testing.
  • Applicable to studies of topoisomerase II inhibition and related pharmacology.
  • Provided with a certificate of analysis to support traceability.