Learn More
Sigma Aldrich Fine Chemicals Biosciences Etoposide, 33419-42-0, MFCD00869325, 100 mg
Supplier:  Sigma Aldrich Fine Chemicals Biosciences E1383100MG
Encompass
Empirical Formula (Hill Notation): C29H32O13, Molecular Weight: 588.56, synthetic, 98.0-105.0%, powder, Synonym: 4′-Demethylepipodophyllotoxin 9-(4,6-O-ethylidene-β-D-glucopyranoside), VP-16-213. Etoposide has been used: to prepare drug stock solution in dimethyl sulfoxide (DMSO) and also to profile and compare the sensitivity of DT40 mutant cells, to incubate cells for cell viability assay, and to treat neuro-2A cells to induce programmed cell death.
Catalog No. NC0916604