Learn More
TARGETMOL CHEMICALS INC Etoposide 500MG
SDP
Catalog No. 502880670
Encompass_Preferred
Also available in 1 mL, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Etoposide (VP-16-213) is a topoisomerase II inhibitor that inhibits DNA synthesis by forming a complex with topoisomerase II and DNA (IC50=60.3 uM). Etoposide has antitumor activity and induces apoptosis and autophagy. Purity 99.95%
Catalog No. 50-288-0670 Supplier TARGETMOL CHEMICALS INC Supplier No. T0132500MG
Also available in 1 mL, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Etoposide (VP-16-213) is a topoisomerase II inhibitor that inhibits DNA synthesis by forming a complex with topoisomerase II and DNA (IC50=60.3 uM). Etoposide has antitumor activity and induces apoptosis and autophagy. Purity 99.95%