Learn More
Cayman Chemical Etoposide, 33419-42-0, 50 mg
SDP
Supplier:  Cayman Chemical 1209250
Encompass_Preferred
Molecular Formula C29H32O13, Purity ≥98%, Formula Weight 588.6, Synonyms: EPE, NSC 141540, VP-16-123; An inhibitor of topoisomerase II (IC50 = 60.3 M); can have much greater potencies when evaluated in cell-based cytotoxicity assays (e.g., IC50 = 5.14 nM for MCF-7 cells); can also inhibit nuclear receptor coactivator 3 (IC50 of 2.48 M).
Catalog No. NC0582295
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.