Learn More
Selleck Chemical LLC Etoposide S1225-10g
SDP
Catalog No. 503108262
Encompass_Preferred
Etoposide is a semisynthetic derivative of podophyllotoxin which inhibits DNA synthesis via topoisomerase II inhibition activity which enhances double-strand and single-strand cleavage of DNA and reversibly inhibits repair by topoisomerase II binding Etoposide induces autophagy mitophagy and apoptosis
Catalog No. 50-310-8262 Supplier Selleck Chemical LLC Supplier No. S122510G
Etoposide is a semisynthetic derivative of podophyllotoxin which inhibits DNA synthesis via topoisomerase II inhibition activity which enhances double-strand and single-strand cleavage of DNA and reversibly inhibits repair by topoisomerase II binding Etoposide induces autophagy mitophagy and apoptosis