Learn More
Cayman Chemical Etoposide-d3, Purity: ≥99% deuterated forms (d1-d3), 1 mg
SDP
Catalog No. 502934014
Encompass_Preferred
An internal standard for the quantification of etoposide by GC- or LC-MS
Catalog No. 50-293-4014 Supplier Cayman Chemical Supplier No. 264881 MG
An internal standard for the quantification of etoposide by GC- or LC-MS