Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Etoposide impurity 1 | 23363-35-1 | MFCD01725594 | 562.52 g/mol | C27H30O13 | 5 MG
SDP

Catalog No. 5000221885
Encompass_Preferred

Etoposide impurity 1 is an analytical impurity standard of the chemotherapeutic agent etoposide, supplied for method development, impurity profiling, and quality control. It is provided as a solid material with defined storage recommendations to maintain stability for analytical use.

  • Analytical reference standard for impurity profiling and method development.
  • White to off-white solid suitable for HPLC and LC-MS analyses.
  • Molecular formula C27H30O13; molecular weight 562.52 g/mol.
  • Available in small pack sizes, including 5 MG, for trace-level studies.
  • Store at 4°C under inert gas; in solvent: -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-21885 Supplier Medchemexpress LLC Supplier No. HY1366695MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Etoposide impurity 1 is an analytical impurity standard of the chemotherapeutic agent etoposide, supplied for method development, impurity profiling, and quality control. It is provided as a solid material with defined storage recommendations to maintain stability for analytical use.

  • Analytical reference standard for impurity profiling and method development.
  • White to off-white solid suitable for HPLC and LC-MS analyses.
  • Molecular formula C27H30O13; molecular weight 562.52 g/mol.
  • Available in small pack sizes, including 5 MG, for trace-level studies.
  • Store at 4°C under inert gas; in solvent: -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.