Learn More
Medchemexpress LLC Etoposide impurity 1 (Standard) | 23363-35-1 | 562.52 | 25 MG
SDP
Catalog No. 5000352334
Encompass_Preferred
Etoposide impurity 1, also known as Lignan P (Standard), is an analytical standard primarily intended for research and analytical applications. It serves as a reference standard, commonly utilized in various methodological research experiments such as High-Performance Liquid Chromatography (HPLC), Gas Chromatography (GC), and Mass Spectrometry (MS).
  • Intended for research and analytical applications
  • Serves as a reference standard
  • Commonly used in HPLC, GC and MS experiments
Catalog No. 50-003-52334 Supplier Medchemexpress LLC Supplier No. 25MGHY136669
Etoposide impurity 1, also known as Lignan P (Standard), is an analytical standard primarily intended for research and analytical applications. It serves as a reference standard, commonly utilized in various methodological research experiments such as High-Performance Liquid Chromatography (HPLC), Gas Chromatography (GC), and Mass Spectrometry (MS).
  • Intended for research and analytical applications
  • Serves as a reference standard
  • Commonly used in HPLC, GC and MS experiments