Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Etoposide impurity 1 (standard) | 23363-35-1 | 562.52 | 50 MG
SDP

Catalog No. 5000348080
Encompass_Preferred

Etoposide impurity 1, also known as Lignan P, is an analytical and reference standard intended strictly for research and analytical applications. It is not for patient use and is commonly employed in qualitative, quantitative, and methodological research experiments.

  • Useful in techniques such as High-Performance Liquid Chromatography (HPLC), Gas Chromatography (GC), and Mass Spectrometry (MS).
  • Molecular weight: 562.52
  • Molecular formula: C27H30O13

Catalog No. 50-003-48080 Supplier Medchemexpress LLC Supplier No. HY13666950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Etoposide impurity 1, also known as Lignan P, is an analytical and reference standard intended strictly for research and analytical applications. It is not for patient use and is commonly employed in qualitative, quantitative, and methodological research experiments.

  • Useful in techniques such as High-Performance Liquid Chromatography (HPLC), Gas Chromatography (GC), and Mass Spectrometry (MS).
  • Molecular weight: 562.52
  • Molecular formula: C27H30O13

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.