Learn More
Medchemexpress LLC Etoposide phosphate | 117091-64-2 | 99.4% | 668.54 | 1 ML
SDP
Catalog No. 5000350970
Encompass_Preferred
Etoposide phosphate is a water-soluble derivative and prodrug of etoposide, functioning as a potent anti-cancer chemotherapy agent and a selective topoisomerase II inhibitor. It prevents re-ligation of DNA strands and induces cell cycle arrest, apoptosis, and autophagy, making it suitable for various research applications.
  • Potent anti-cancer chemotherapy agent
  • Selective topoisomerase II inhibitor
  • Prevents re-ligation of DNA strands
  • Induces cell cycle arrest, apoptosis, and autophagy
  • Water-soluble derivative and probable prodrug of etoposide
  • Inhibits HCT116 cell growth in a dose-dependent manner
Catalog No. 50-003-50970 Supplier Medchemexpress LLC Supplier No. HY1363010MM/1ML
Etoposide phosphate is a water-soluble derivative and prodrug of etoposide, functioning as a potent anti-cancer chemotherapy agent and a selective topoisomerase II inhibitor. It prevents re-ligation of DNA strands and induces cell cycle arrest, apoptosis, and autophagy, making it suitable for various research applications.
  • Potent anti-cancer chemotherapy agent
  • Selective topoisomerase II inhibitor
  • Prevents re-ligation of DNA strands
  • Induces cell cycle arrest, apoptosis, and autophagy
  • Water-soluble derivative and probable prodrug of etoposide
  • Inhibits HCT116 cell growth in a dose-dependent manner