Learn More
Medchemexpress LLC Etoposide (Standard) | 33419-42-0 | 99.6% | 588.56 | 100 MG
SDP
Catalog No. 5000349570
Encompass_Preferred
Etoposide (Standard) is an analytical standard of Etoposide, intended for research and analytical applications. Etoposide is an anti-cancer chemotherapy agent that inhibits topoisomerase II, thereby stopping DNA replication. It induces cell cycle arrest, apoptosis, and autophagy.
  • Serves as a reference standard for assays
  • Suitable for qualitative, quantitative, and methodological research experiments (e.g., HPLC, GC, MS)
  • Inhibits topoisomerase II
  • Induces cell cycle arrest, apoptosis, and autophagy
Catalog No. 50-003-49570 Supplier Medchemexpress LLC Supplier No. HY13629R100MG
Etoposide (Standard) is an analytical standard of Etoposide, intended for research and analytical applications. Etoposide is an anti-cancer chemotherapy agent that inhibits topoisomerase II, thereby stopping DNA replication. It induces cell cycle arrest, apoptosis, and autophagy.
  • Serves as a reference standard for assays
  • Suitable for qualitative, quantitative, and methodological research experiments (e.g., HPLC, GC, MS)
  • Inhibits topoisomerase II
  • Induces cell cycle arrest, apoptosis, and autophagy