Learn More
Selleck Chemical LLC Etoposide 500mg 33419-42-0 VP-16, VP-16213
SDP
Catalog No. 507192
Encompass_Preferred
Etoposide (VP-16, VP-16213) is a semisynthetic derivative of podophyllotoxin, which inhibits DNA synthesis via topoisomerase II inhibition activity. Etoposide induces autophagy, mitophagy and apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-719-2 Supplier Selleck Chemical LLC Supplier No. S1225500MG
Add to Cart
Add to Cart
Etoposide (VP-16, VP-16213) is a semisynthetic derivative of podophyllotoxin, which inhibits DNA synthesis via topoisomerase II inhibition activity. Etoposide induces autophagy, mitophagy and apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.