Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Etoricoxib

Catalog No. AC463380010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC463380010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC463380010 Supplier Thermo Scientific Chemicals Supplier No. 463380010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 202409-33-4 | C18H15ClN2O2S | 358.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 202409-33-4
Molecular Formula C18H15ClN2O2S
Molecular Weight (g/mol) 358.84
InChI Key MNJVRJDLRVPLFE-UHFFFAOYSA-N
IUPAC Name 5-chloro-3-(4-methanesulfonylphenyl)-6'-methyl-2,3'-bipyridine
SMILES CC1=CC=C(C=N1)C1=NC=C(Cl)C=C1C1=CC=C(C=C1)S(C)(=O)=O

Specifications

Melting Point 134.0°C to 138.0°C
Quantity 1 g
Formula Weight 358.84
Chemical Name or Material Etoricoxib
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Harmful if swallowed.

Fatal in contact with skin.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Wear protective gloves/protective clothing.

Do not get in eyes, on skin, or on clothing.

IF ON SKIN: Gently wash with plenty of soap and water.

Immediately call a POISON CENTER or doctor/physician.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.