Learn More
AdipoGen Etoricoxib (25 mg)
SDP
Supplier:  AdipoGen CDXE0092M025
Encompass_Preferred
Chemical. CAS: 202409-33-4. Formula: C18H15ClN2O2S. MW: 358.84. Etoricoxib is a non-steroidal anti-inflammatory drug (NSAID) and a potent cyclooxygenase-2 (COX-2) inhibitor. This dipyridinyl compound demonstrates high in vitro and ex vivo selectivity for COX-2 over COX-1 in several assays. Used to treat patients suffering from osteoarthritis and rheumatoid arthritis. Might have some chemopreventive properties.
Catalog No. 50-205-4413