Learn More
Agilent Technologies eTox Red, Cell Viability Stain, Liquid
Catalog No. NC3737481
Encompass
eTox Red is a fluorescent dye designed for the selective labeling of dead cells in live-cell imaging experiments. This reagent is incapable of crossing the intact membranes of healthy, viable cells, but it readily gains entry to the interior of dead cells through compromised membrane structures, where it binds to DNA to emit a red fluorescent signal. It is intended for research use only applications within the xCELLigence RTCA eSight platform.
  • Selectively stains the nuclei of dead cells
  • Ready to use formulation for live-cell imaging
  • Compatible with the xCELLigence RTCA eSight system
  • Liquid physical state
  • Miscible with water
Catalog No. NC3737481 Supplier Agilent Technologies Supplier No. 8711009
eTox Red is a fluorescent dye designed for the selective labeling of dead cells in live-cell imaging experiments. This reagent is incapable of crossing the intact membranes of healthy, viable cells, but it readily gains entry to the interior of dead cells through compromised membrane structures, where it binds to DNA to emit a red fluorescent signal. It is intended for research use only applications within the xCELLigence RTCA eSight platform.
  • Selectively stains the nuclei of dead cells
  • Ready to use formulation for live-cell imaging
  • Compatible with the xCELLigence RTCA eSight system
  • Liquid physical state
  • Miscible with water