Learn More
AdipoGen Etoxazole
SDP
Supplier:  AdipoGen CDXE0064M100
Encompass_Preferred
Chemical. CAS 153233-91-1. Formula C21H23F2NO2. MW 359.4. Synthetic. Acaricide/Insecticide. Mode of action is via chitin biosynthesis inhibition for the control of mites. Compound can be used as analytical reference material.
Catalog No. 50-256-0982