Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Etoxazole-d5 | 153233-91-1 | 98.0% | 364.44 g/mol | C21H18D5F2NO2 | 5 MG
SDP

Catalog No. 5000210479
Encompass_Preferred

Etoxazole-d5 is a deuterium-labeled research standard used as a tracer and as an internal standard for quantitative analyses by NMR, GC-MS, or LC-MS.

  • Deuterium-labeled isotopologue used as a tracer or internal standard.
  • Purity 98.0%.
  • Molecular formula C21H18D5F2NO2; molecular weight 364.44 g/mol.
  • White to off-white solid appearance.
  • Storage: powder -20 °C (3 years) or 4 °C (2 years); in solvent -80 °C (6 months) or -20 °C (1 month).
  • Available in small research sizes, including 5 mg.

Catalog No. 50-002-10479 Supplier Medchemexpress LLC Supplier No. HYW059342S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Etoxazole-d5 is a deuterium-labeled research standard used as a tracer and as an internal standard for quantitative analyses by NMR, GC-MS, or LC-MS.

  • Deuterium-labeled isotopologue used as a tracer or internal standard.
  • Purity 98.0%.
  • Molecular formula C21H18D5F2NO2; molecular weight 364.44 g/mol.
  • White to off-white solid appearance.
  • Storage: powder -20 °C (3 years) or 4 °C (2 years); in solvent -80 °C (6 months) or -20 °C (1 month).
  • Available in small research sizes, including 5 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.