Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Etoxazole-d5 | 98.0% | 364.44 g/mol | C21H18D5F2NO2 | 1 MG
SDP

Catalog No. 5000209987
Encompass_Preferred

Etoxazole-d5 is the deuterium-labeled form of etoxazole intended for use as an isotope-labeled reference in analytical and research applications. It is used as a tracer or internal standard for quantitation and environmental or metabolic studies.

  • Isotope-labeled reference standard for analytical quantitation.
  • Deuterated (D5) analog of etoxazole.
  • Molecular weight: 364.44 g/mol.
  • Chemical formula: C21H18D5F2NO2.
  • Available in small-quantity packs suitable for standards (e.g., 1 mg).

Catalog No. 50-002-09987 Supplier Medchemexpress LLC Supplier No. HYW059342S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Etoxazole-d5 is the deuterium-labeled form of etoxazole intended for use as an isotope-labeled reference in analytical and research applications. It is used as a tracer or internal standard for quantitation and environmental or metabolic studies.

  • Isotope-labeled reference standard for analytical quantitation.
  • Deuterated (D5) analog of etoxazole.
  • Molecular weight: 364.44 g/mol.
  • Chemical formula: C21H18D5F2NO2.
  • Available in small-quantity packs suitable for standards (e.g., 1 mg).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.