Learn More
ASTREA BIOSEPARATIONS US INC EtoxiClear® Pre-packed Column, 1 x 5 mL
Catalog No. NC2832116
Encompass
EtoxiClear® 5 mL Pre-packed Columns are an essential tool for efficient affinity endotoxin depletion and removal. Easy-to-use and perfectly suited to single-use applications, these small-scale columns are compatible with automated systems or manual operations, making them a versatile choice for screening and process development.
Catalog No. NC2832116 Supplier ASTREA BIOSEPARATIONS US INC Supplier No. 6747
EtoxiClear® 5 mL Pre-packed Columns are an essential tool for efficient affinity endotoxin depletion and removal. Easy-to-use and perfectly suited to single-use applications, these small-scale columns are compatible with automated systems or manual operations, making them a versatile choice for screening and process development.